Storage boxes, LIGHT BOX
Supplier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2291 2292 2293 2294 2295 2296 2299 2298 2288
MULC2291EA 34.1 GBP
MULC2291 MULC2292 MULC2293 MULC2294 MULC2295 MULC2296 MULC2299 MULC2298 MULC2288
Storage boxes, LIGHT BOX
Boxes Storage Boxes
Made of polypropylene.
  • Totally transparent
  • Slip lid with secure clip locking system
  • Shock resistant
These storage boxes are perfectly suited to the storage and organisation of differently sized products. The contents can be easily identified through the transparent material.