Absorption granules - Universal coarse grain
Supplier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9956 18468
MSGA9956EA 41.8 GBP
MSGA9956 MSGA18468
Absorption granules - Universal coarse grain
Spill Control Materials
Absorption granules for oil, alkalis, grease, motor fuels, varnishes.
  • For outdoor and indoor use
  • Officially tested and approved for use on traffic areas
  • 20 kg sack binds up to 16 litres of heating oil
  • Suitable for solid surfaces
  • Even with total saturation of the granules, anti-slipping is guaranteed