GAP Hazardous Material Work Stations
Supplier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40566901 40563901 40564901 40567901 40569901 40570901 40573901 40572901 40582901 40581901 40586901 40587901 41044901
MSGA40566901EA 6300 GBP
MSGA40566901 MSGA40563901 MSGA40564901 MSGA40567901 MSGA40569901 MSGA40570901 MSGA40573901 MSGA40572901 MSGA40582901 MSGA40581901 MSGA40586901 MSGA40587901 MSGA41044901
GAP Hazardous Material Work Stations
Fume Hoods and Enclosures Chemical Fume Hoods
Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.
  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and
More Product Information