Safety Storage Cabinets, Type 90, V-CLASSIC-90 MULTIRISK V90.196.060.MV.2VD:0012
Supplier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40285005 40498 40627
MSGA40285005EA 6460 GBP
MSGA40285005 MSGA40498 MSGA40627
Safety Storage Cabinets, Type 90, V-CLASSIC-90 MULTIRISK V90.196.060.MV.2VD:0012
Furniture Storage Cabinets
Safe, flexible, and energetically sustainable storage of various hazardous materials in just one cabinet.
  • Special fire resistant construction
  • Locking
  • Comfortable overview and access
  • Metal-free storage levels
  • Optimal integration and use of space
With a cabinet width of only 60 cm, the MULTIRISK cabinet offers a high storage capacity (per storage level 14×1 L laboratory glass bottles or 4×2.5 L and 3×1 L laboratory glass bottles), as the cabinet with its depth of 86 cm makes perfect use of the available space next to laboratory fume cupboards. TheMore Product Information