Safety storage cabinets, CX-Classic-G
Supplier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32925041 32925050
MSGA32925041EA 5110 GBP
MSGA32925041 MSGA32925050
Safety storage cabinets, CX-Classic-G
Furniture Storage Cabinets
Fireproof safety cabinets for safe and approved storage of water pollutants, non flammable liquids and toxics in working areas according to TRGS 510.
  • Robust construction and longevity- high-quality, scratch-resistant outer body with powder-coated textured surface
  • Doors lockable with cylinder locking (integrated in the turning handle)
  • Adjusting aids to compensate for uneven floor
Cabinet body colour: anthracite grey RAL 7016.
Cabinet features with perforated insert useable as storage area. It consists of bottom collecting sump with galvanised orMore Product Information