Shelving systems for hazardous substances, RWW

Supplier: asecos
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

27829 27830 27831 27832 27833 27834 27835 27836
MSGA27829EA 730 GBP
MSGA27829 MSGA27830 MSGA27831 MSGA27832 MSGA27833 MSGA27834 MSGA27835 MSGA27836
Shelving systems for hazardous substances, RWW
Furniture Shelves
Sturdy construction racks with four galvanized steel sumps shelves as storage levels and galvanized rack frames. Ideal for safe and approved storage of water pollutants.

  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually

Certifications: TÜV approved.

Delivery information: Flat-pack delivery with assembly instructions.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR