Betaine hydrochloride

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M87285084
MOLEM87285084EA 0 GBP
MOLEM87285084
Betaine hydrochloride
Betaine hydrochloride
Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
EINECS: 209-683-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR