5-Methoxy-m-cresol

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

M87126158
MOLEM87126158EA 0 GBP
MOLEM87126158
5-Methoxy-m-cresol
5-Methoxy-m-cresol
Formula: C₈H₁₀O₂
MDL Number: MFCD00059261
CAS Number: 3209-13-0
EINECS: 221-716-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR