Tetraethylammonium hydroxide 40% in aqueous solution

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tetrylammonium hydroxide

M86474667
MOLEM86474667EA 0 GBP
MOLEM86474667
Tetraethylammonium hydroxide 40% in aqueous solution
Tetraethylammonium hydroxide
Formula: C₈H₂₁NO
MW: 147.26 g/mol
Density: 0.865
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 3267
ADR: 8 II
Merck Index: 12,09342

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR