2-Amino-2',5-dichlorobenzophenone

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone

M83820930
MOLEM83820930EA 0 GBP
MOLEM83820930
2-Amino-2',5-dichlorobenzophenone
2-Amino-2',5-dichlorobenzophenone
Formula: C₁₃H₉Cl₂NO
MW: 266.13 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 88…91 °C
MDL Number: MFCD00007840
CAS Number: 2958-36-3
EINECS: 220-985-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR