Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5′-AMP-Na2 hydrate

M79277079
MOLEM79277079EA 0 GBP
MOLEM79277079
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Formula: C₁₀H₁₂N₅Na₂O₇P·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00065023
CAS Number: 4578-31-8
EINECS: 224-961-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR