Estradiol

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M59540594
MOLEM59540594EA 0 GBP
MOLEM59540594
Estradiol
Estradiol
Formula: C₁₈H₂₄O₂
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR