Sodium amide

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

M58630340
MOLEM58630340EA 0 GBP
MOLEM58630340
Sodium amide
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011117
CAS Number: 7782-92-5
EINECS: 231-971-0
UN: 1390
ADR: 4.3,II
Merck Index: 12,08717

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR