Lithium hydroxide monohydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

M49289225
MOLEM49289225EA 0 GBP
MOLEM49289225
Lithium hydroxide monohydrate
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR