2',5'-Dihydroxyacetophenone

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone

M45596894
MOLEM45596894EA 0 GBP
MOLEM45596894
2',5'-Dihydroxyacetophenone
2',5'-Dihydroxyacetophenone
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 199…202 °C
MDL Number: MFCD00002343
CAS Number: 490-78-8
EINECS: 207-716-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR