Thymidine (2'-deoxythymidine)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

M40829422
MOLEM40829422EA 0 GBP
MOLEM40829422
Thymidine (2'-deoxythymidine)
Thymidine (2'-Deoxythymidine)
Formula: C₁₀H₁₄N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS Number: 50-89-5
EINECS: 200-070-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR