3,4-Difluoroaniline

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

M39226244
MOLEM39226244EA 0 GBP
MOLEM39226244
3,4-Difluoroaniline
3,4-Difluoroaniline
Formula: C₆H₅F₂N
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 22 °C
Density: 1.302 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR