3-Chlorocinnamic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(3-Chlorophenyl)acrylic acid, (2E)-3-(3-Chlorophenyl)acrylic acid

M22709700
MOLEM22709700EA 0 GBP
MOLEM22709700
3-Chlorocinnamic acid
3-Chlorocinnamic acid
Formula: C₉H₇ClO₂
MW: 182.61 g/mol
MDL Number: MFCD00004384
CAS Number: 1866-38-2
EINECS: 217-478-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR