2,4-Dihydroxybenzaldehyde

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

78232322-25G
MOLE78232322-25GEA 0 GBP
MOLE78232322-25G
2,4-Dihydroxybenzaldehyde
2,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 132…133 °C
Density: 1.448 g/cm³ (–93 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 12,08324

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR