4,4'-Dimethoxytrityl chloride

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride

65276715-100G
MOLE65276715-100GEA 0 GBP
MOLE65276715-100G
4,4'-Dimethoxytrityl chloride
4,4'-Dimethoxytrityl chloride
Formula: C₂₁H₁₉ClO₂
MDL Number: MFCD00008409
CAS Number: 40615-36-9
EINECS: 255-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR