Mercury(II) sulphate 200 g/l in aqueous solution

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Mercury sulphate, Mercuric sulphate

61441891-1L
MOLE61441891-1LEA 0 GBP
MOLE61441891-1L
Mercury(II) sulphate 200 g/l in aqueous solution
Mercury (II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 2922
ADR: 8 II
Merck Index: 13,05914

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR