Tetrabutylammonium dihydrogen phosphate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

46939015-5G
MOLE46939015-5GEA 0 GBP
MOLE46939015-5G
Tetrabutylammonium dihydrogen phosphate
Tetrabutylammonium dihydrogen phosphate
Formula: C₁₆H₃₈NO₄P
MDL Number: MFCD00064526
CAS Number: 5574-97-0
EINECS: 226-947-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR