2-Naphthaldehyde

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: β-Naphthaldehyde

40933039-25G
MOLE40933039-25GEA 0 GBP
MOLE40933039-25G
2-Naphthaldehyde
2-Naphthaldehyde
Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160 °C (13 mmHg)
Melting Pt: 57…60 °C
Density: 1.0775 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR