2,6-Dichloropyridine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

34574238-100G
MOLE34574238-100GEA 0 GBP
MOLE34574238-100G
2,6-Dichloropyridine
2,6-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR