1,10-Phenanthroline hydrochloride monohydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

28352631-25G
MOLE28352631-25GEA 0 GBP
MOLE28352631-25G
1,10-Phenanthroline hydrochloride monohydrate
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
Melting Pt: 224…226 °C
Density: 1.385 g/cm³ (–100.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR