5-(4-(Dimethylamino)benzylidene)rhodanine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

24067530-50G
MOLE24067530-50GEA 0 GBP
MOLE24067530-50G
5-(4-(Dimethylamino)benzylidene)rhodanine
5-(4-(Dimethylamino)benzylidene)rhodanine
Formula: C₁₂H₁₂N₂OS₂
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR