2,6-Difluorobenzaldehyde

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

23482755-5G
MOLE23482755-5GEA 0 GBP
MOLE23482755-5G
2,6-Difluorobenzaldehyde
2,6-Difluorobenzaldehyde
Formula: C₇H₄F₂O
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 15…17 °C
Density: 1,31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010293
CAS Number: 437-81-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR