Picolinic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid

23001678-100G
MOLE23001678-100GEA 0 GBP
MOLE23001678-100G
Picolinic acid
Picolinic acid
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 134…137 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR