Magnesium sulphate heptahydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22772499
MOLE22772499EA 0 GBP
MOLE22772499
Magnesium sulphate heptahydrate
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR