MOPS-Na

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22144595-100G
MOLE22144595-100GEA 0 GBP
MOLE22144595-100G
MOPS-Na
MOPS-Na
Formula: C₇H₁₄NNaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00064350
CAS Number: 71119-22-7
EINECS: 428-420-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR