5-Methylphenazinium methyl sulphate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

20097670-25G
MOLE20097670-25GEA 0 GBP
MOLE20097670-25G
5-Methylphenazinium methyl sulphate
5-Methylphenazinium methyl sulphate
Formula: C₁₄H₁₄N₂O₄S
Melting Pt: 198 °C
Storage Temperature: Freezer
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR