Ammonium heptamolybdate tetrahydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

19757224-1KG
MOLE19757224-1KGEA 0 GBP
MOLE19757224-1KG
Ammonium heptamolybdate tetrahydrate
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR