1,1,3,3-Tetramethylguanidine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

17115209-100G
MOLE17115209-100GEA 0 GBP
MOLE17115209-100G
1,1,3,3-Tetramethylguanidine
1,1,3,3-Tetramethylguanidine
Formula: C₅H₁₃N₃
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR