n-Undecyl-β-D-maltoside

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

16709684-1G
MOLE16709684-1GEA 0 GBP
MOLE16709684-1G
n-Undecyl-β-D-maltoside
n-Undecyl-β-D-maltoside
Formula: C₂₃H₄₄O₁₁
CAS Number: 170552-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR