2-Chloropyrimidine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

16639234-25G
MOLE16639234-25GEA 0 GBP
MOLE16639234-25G
2-Chloropyrimidine
2-Chloropyrimidine
Formula: C₄H₃ClN₂
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65…66 °C
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR