Anthranilic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

15557294-1KG
MOLE15557294-1KGEA 0 GBP
MOLE15557294-1KG
Anthranilic acid
Anthranilic acid
Formula: C₇H₇NO₂
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 144 °C
Density: 1.362 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007712
CAS Number: 118-92-3
EINECS: 204-287-5
Merck Index: 13,00421

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR