15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 15-Crown-5, 1,4,7,10,13-Pentaoxacyclopentadecane

10920590-25G
MOLE10920590-25GEA 0 GBP
MOLE10920590-25G
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
15-Crown-5 (1,4,7,10,13-Pentaoxacyclopentadecane, Lead ionophore V)
Formula: C₁₀H₂₀O₅
MW: 220.27 g/mol
Boiling Pt: 351
Melting Pt: –20 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005110
CAS Number: 33100-27-5
EINECS: 251-379-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR