Digital timer, two channels, 580CE
Supplier: MOINEAU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0580CE
MOIN0580CEEA 120 GBP
MOIN0580CE
Digital timer, two channels, 580CE
Timers Digital Timers
Digital timer with two channels and two display lines.
  • Timing capacity: 100 hours
  • Counts down and up
  • Memory for both channels
  • Adjustable alarm volume
  • Acoustic and visual alarm
Weight: 85 g
Delivery information: Supplied with stand, clamp and magnet. Batteries (2× AAA) need to be ordered separately.