Flip-top dilution bottles
Supplier: 3M Food Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FTBFD9060 FTBPW9060
MMMPFTBFD9060EA 87.8 GBP
MMMPFTBFD9060 MMMPFTBPW9060
Flip-top dilution bottles
Bottles Dilution Bottles
The 3M™ Flip-top dilution bottle is a leak-resistant plastic cylinder with a flip-open lid that seals securely for safe shipping and handling.
  • Hinged lid stays open for easy sample additions
  • Ready for use with no additional blending required
  • Generous headspace and bottle opening make sample addition easy
  • Leak resistant vial helps prevent leakage during shipping
  • 2 year shelf life from date of manufacture provides long-term readiness
3M™ Flip-top dilution bottles are ideal diluents for enumeration of microorganisms from food, beverage, and environmentalMore Product Information