Spore strips, Attest™
Supplier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M1292S M1292E
MMMEM1292SEA 1630 GBP
MMMEM1292S MMMEM1292E
Spore strips, Attest™
Sterilisation Indicators
Biological indicators provide the best assurance of sterility by challenging the steriliser with quantifiable, highly resistant spores. After sterilisation, the vial is 'crushed' to join the growth media with the processed spore strip. A colour change to yellow indicates surviving spores and a positive result. Biological indicators are used for load control monitoring which is the process by which a load is monitored and released based on the result of a biological indicator (BI) in a test pack.
  • The self-contained (BI) for 121 °C gravity/132 °C vacuum-assisted steam cycles consist of a
More Product Information