4-Station battery charger kit TR-344E, Versaflo™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TR344UK
MMMATR344UKEA 0 GBP
MMMATR344UK
4-Station battery charger kit TR-344E, Versaflo™
Batteries
The Versaflo™ 4-station Battery Charger Kit TR-344E is designed for use with the Versaflo™ Powered Air Turbo TR-300+.
  • 4-station battery charger
  • CE approved
  • Reverts to trickle charge when battery is full so it can remain plugged-in on standby
The Versaflo™ 4-Station Battery Charger TR-344E is designed for use with the Versaflo™ Powered Air Turbo TR-300+. This charger kit features integrated LEDs which indicate the charge status of the battery and alerts users if a fault occurs. It has four stations, making it ideal for charging multiple batteries at a time on-site.