Powered air respirator airflow indicator TR-970
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TR-970
MMMATR-970EA 38.6 GBP
MMMATR-970
Powered air respirator airflow indicator TR-970
Respirators Respiratory Systems
The TR-970 Air Flow Indicator is used to confirm that the manufacturer's minimum design flow rate of the Versaflo™ TR-300 Series Powered Air Turbo is exceeded.
  • Designed to enable testing at various elevations and temperatures within the operational range
  • Easy to use
Order Now
SPECIFICATIONS
Environmentally Preferable