Powered air respirator filter cover for Versaflo™ TR-6580E, Versaflo™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TR-6500FC
MMMATR-6500FCEA 21.8 GBP
MMMATR-6500FC
Powered air respirator filter cover for Versaflo™ TR-6580E, Versaflo™
Respirators Respiratory Systems
The Versaflo™ Filter Cover TR-6500FC is a filter cover that helps to protect Versaflo™ TR-6580E filters from physical damage.
  • Protects 3M™ Versaflo™ TR-6580E filter from physical damage
  • Can be cleaned and reused
This filter cover safeguards the filter and enables users to wear Versaflo™ Powered Air Turbo through decontamination showers. It can hold a Versaflo™ Spark Arrestor TR-662, or Versaflo™ Prefilter TR-6600 in place for additional protection from larger particles or sparks. The filter cover can be cleaned for reuse.