Supplied air system, V-series, Versaflo™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S212
MMMAS212EA 444 GBP
MMMAS212
Supplied air system, V-series, Versaflo™
Respirators Respiratory Systems
Regulator Assembly S-212 is a replacement part for use with Versaflo™ Supplied Air System S-200+.
  • Spare or replacement part
  • Designed for use with 3M™ Versaflo™ Supplied Air System S-200+
  • CE approved
Prolong the service life of the 3M™ Versaflo™ Supplied Air System S-200+ with 3M™ Regulator Assembly, S-212. This spare regulator is specially designed for S-200+ systems.