Powered air respirator over-visors, Versaflo™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M-967
MMMAM-967EA 195 GBP
MMMAM-967
Powered air respirator over-visors, Versaflo™
Respirators Respiratory Systems
The Versaflo™ Over-Visor, gold, IR/UV shade 5, M-967, is an over-visor to protect against UV/IR, metal sparks, and splashes.
  • Gold IR/UV over-visor
  • For use with Versaflo™ Over-visor Frame, M-961
Get IR and UV protection when needed with the Versaflo™ Over-visor, Gold, IR/UV Shade 5, M-967. This over-visor measures 500×250 mm and protects users from UV as well as IR. It’s designed for use with 3M™ Versaflo™ Over-visor Frame, M-961.