Secure Click™ FF-800 Full Facepiece Reusable Respirators
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FF-801 FF-802 FF-803
MMMAFF-801EA 345 GBP
MMMAFF-801 MMMAFF-802 MMMAFF-803
Secure Click™ FF-800 Full Facepiece Reusable Respirators
Respirators Reusable Respirators Full Face Respirators
The 3M™ Secure Click™ full facepiece reusable respirator FF-800 series helps provide protection against certain airborne contaminants. For use with 3M™ Secure Click™ particulate filters and 3M™ Secure Click™ multi-gas/vapor cartridges.
  • Quick, simple cartridge/filter connection. Simply align the connections and push until you hear a click
  • Quick, easy one-touch positive pressure user seal check
  • Speaking diaphragm is designed to help provide easy communication
  • Scotchgard™ Protector on lens for improved paint and stain resistance
The FF-800 seriesMore Product Information