Disposable earplugs, E-A-R™ Push-Ins™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EX01020 EX01021
111-1798EA 123 GBP
111-1798 MMMAEX01021
Disposable earplugs, E-A-R™ Push-Ins™
Earplugs Disposable Earplugs
These pod earplugs are very easy to insert since there is no roll needed, users can just use the soft flexible grip to push the earplug into position. As the user therefore does not have to touch the foam during fitting it is one of the most hygienic types of earplug.
  • Flexible stem makes insertion and removal cleaner and easier
  • E-A-Rform foam softens with body heat to create an individualised fit that gets more comfortable with longer wear
  • Low pressure seal, available with or without cord
Suitable for high noise exposure levels and particularly forMore Product Information