General-purpose conformable duct tapes, 2903, Scotch®
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2903-BL 2903-SV
MMMA2903-BLEA 259 GBP
MMMA2903-BL MMMA2903-SV
General-purpose conformable duct tapes, 2903, Scotch®
Tapes
Scotch® general purpose duct tape 2903 is a versatile, polyethylene film on canvas fabric with a synthetic rubber finish.
  • Good initial tack to a wide variety of surfaces
  • Highly conformable
  • High tensile strength, ideal for bundling
  • Tears easily by hand in both directions
  • Individually wrapped for easy identification and to keep the roll clean until ready for use
  • Tensile strength: 100 [Newton/25 mm]
  • PE coated cloth scrim back material
General-purpose, conformable duct tape is ideal for general maintenance, bundling, wrapping, attaching, sealing, andMore Product Information