Value duct tape, 1900
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1900S50 1900SI50
MMMA1900S50EA 12.5 GBP
MMMA1900S50 MMMA1900SI50
Value duct tape, 1900
Tapes
3M™ value duct tape 1900 is a cost-effective duct tape for general utility use and lighter duty non-critical applications. Ideal for general applications including light-duty sealing, bundling, marking, and temporary repairs.
  • Polyethylene backing over cloth scrim resists moisture for reliable sealing performance in moist or humid environments
  • Conforms to irregular surfaces without lifting
  • Easy to unwind and easy to tear
This tape features a synthetic rubber adhesive that sticks on contact to smooth metal, glass, plastic, sealed concrete, and more. TheMore Product Information