High content cell imaging system, micro confocal, ImageXpress
Supplier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MICROCONFOCAL 5039731 5040287 5041314 5041309 5041310 5041312 MICROCONFO_REF
735-0364EA 0 GBP
735-0364 MLDV5039731 MLDV5040287 MLDV5041314 MLDV5041309 MLDV5041310 MLDV5041312 MLDVMICROCONFO_REF
High content cell imaging system, micro confocal, ImageXpress
Microscopes
The ImageXpress® Micro-confocal high content imaging system helps to expand the boundaries of research with a combination of speed, sensitivity and flexibility, giving you higher quality images, at a faster throughput, with more powerful analysis.
  • Quickly and easily switch between widefield and several confocal disc configurations, including an exclusive slit option, to capture images at high throughput speeds
  • Image more than 160000 wells per day in confocal mode
  • Excellent image quality via the unique optical path technology
  • Large field of view enables
More Product Information